Please login to your online account to display your discounted pricing

anti-ILT5/CD85a, PE, Clone: 222821, R&D Systems

$129.05 - $363.69

Specifications

Antigen ILT5/CD85a
Clone 222821
Cross Reactivity Human
Format Purified
Host Species Mouse
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

FAB1806P025

 
r & d systems
RDSFAB1806P025
PE 25 Tests Each for $129.05

FAB1806P100

 
r & d systems
RDSFAB1806P100
PE 100 Tests Each for $363.69
Description & Specifications

Specifications

Antigen ILT5/CD85a
Clone 222821
Cross Reactivity Human
Format Purified
Host Species Mouse
Regulatory Status RUO
Immunogen Mouse myeloma cell line NS0-derived recombinant human ILT5/CD85a. Gly24-Glu443
Isotype IgG2a
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Purification Method Protein A or G purified from hybridoma culture supernatant

Applications:

Flow Cytometry