Please login to your online account to display your discounted pricing

anti-ILT5/CD85a, APC, Clone: 222821, R&D Systems


Manufacturer: r & d systems  RDSFAB1806A025

 View more versions of this product

Catalog No. FAB1806A025

  • / Each
 

Description & Specifications

Specifications

Antigen ILT5/CD85a
Clone 222821
Conjugate Allophycocyanin
Cross Reactivity Human
Format Purified
Host Species Murine
Immunogen Mouse myeloma cell line NS0-derived recombinant human ILT5/CD85a. Gly24-Glu443
Isotype IgG2a
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human