Please login to your online account to display your discounted pricing

anti-ILT4/CD85d, PerCP, Clone: 287219, R&D Systems


Manufacturer: r & d systems  RDSFAB2078C025

 View more versions of this product

Catalog No. FAB2078C025

  • / Each
 

Description & Specifications

Specifications

Antigen ILT4/CD85d
Immunogen Mouse myeloma cell line NS0-derived recombinant human ILT4/CD85d. Gly24-His458
Format Purified
Host Species Mouse
Isotype IgG2a
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone 287219
Conjugate PerCP
Cross Reactivity Human