Please login to your online account to display your discounted pricing

anti-ILT4/CD85d, PerCP, Clone: 287219, R&D Systems

$126.32 - $394.29

Specifications

Antigen ILT4/CD85d
Clone 287219
Cross Reactivity Human
Format Purified
Host Species Mouse
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

FAB2078C100

 
r & d systems
RDSFAB2078C100
PerCP 100 Tests Each for $394.29

FAB2078C025

 
r & d systems
RDSFAB2078C025
PerCP 25 Tests Each for $126.32
Description & Specifications

Specifications

Antigen ILT4/CD85d
Clone 287219
Cross Reactivity Human
Format Purified
Host Species Mouse
Regulatory Status RUO
Immunogen Mouse myeloma cell line NS0-derived recombinant human ILT4/CD85d. Gly24-His458
Isotype IgG2a
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Purification Method Protein A or G purified from hybridoma culture supernatant

Applications:

Flow Cytometry