Please login to your online account to display your discounted pricing

anti-ILT4/CD85d, PerCP, Clone: 287219, R&D Systems


Manufacturer: r & d systems  RDSFAB2078C100

 View more versions of this product

Catalog No. FAB2078C100

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Host Species Mouse
Antigen ILT4/CD85d
Isotype IgG2a
Cross Reactivity Human
Conjugate PerCP
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone 287219
Immunogen Mouse myeloma cell line NS0-derived recombinant human ILT4/CD85d. Gly24-His458
Quantity 100 Tests
Format Purified
Purification Method Protein A or G purified from hybridoma culture supernatant