Please login to your online account to display your discounted pricing

anti-ILT4/CD85d, PE, Clone: 287219, R&D Systems


Manufacturer: r & d systems  RDSFAB2078P100

 View more versions of this product

Catalog No. FAB2078P100

  • / Each
 

Description & Specifications

Specifications

Antigen ILT4/CD85d
Regulatory Status RUO
Cross Reactivity Human
Format Purified
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human ILT4. Gly24-His458
Isotype IgG2a
Quantity 100 Tests
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone 287219
Conjugate PE
Purification Method Protein A or G purified from hybridoma culture supernatant