Please login to your online account to display your discounted pricing

anti-ILT4/CD85d, PE, Clone: 287219, R&D Systems


Manufacturer: r & d systems  RDSFAB2078P025

 View more versions of this product

Catalog No. FAB2078P025

  • / Each
 

Description & Specifications

Specifications

Antigen ILT4/CD85d
Clone 287219
Conjugate PE
Cross Reactivity Human
Format Purified
Host Species Murine
Immunogen Mouse myeloma cell line NS0-derived recombinant human ILT4. Gly24-His458
Isotype IgG2a
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human