Please login to your online account to display your discounted pricing

anti-ILT4/CD85d, Fluorescein, Clone: 287219, R&D Systems


Manufacturer: r & d systems  RDSFAB2078F025

 View more versions of this product

Catalog No. FAB2078F025

  • / Each
 

Description & Specifications

Specifications

Format Purified
Isotype IgG2a
Quantity 25 Tests
Clone 287219
Cross Reactivity Human
Antigen ILT4/CD85d
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human ILT4. Gly24-His458
Purification Method Protein A or G purified from hybridoma culture supernatant
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate Fluorescein