Please login to your online account to display your discounted pricing

anti-ILT4/CD85d, Fluorescein, Clone: 287219, R&D Systems


Manufacturer: r & d systems  RDSFAB2078F100

 View more versions of this product

Catalog No. FAB2078F100

  • / Each
 

Description & Specifications

Specifications

Format Purified
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human ILT4. Gly24-His458
Antigen ILT4/CD85d
Cross Reactivity Human
Conjugate Fluorescein
Isotype IgG2a
Primary or Secondary Primary
Quantity 100 Tests
Clone 287219
Regulatory Status RUO
Purification Method Protein A or G purified from hybridoma culture supernatant
Monoclonal or Polyclonal Monoclonal