Please login to your online account to display your discounted pricing

anti-ILT4/CD85d, APC, Clone: 287219, R&D Systems


Manufacturer: r & d systems  RDSFAB2078A025

 View more versions of this product

Catalog No. FAB2078A025

  • / Each
 

Description & Specifications

Specifications

Format Purified
Isotype IgG2a
Regulatory Status RUO
Primary or Secondary Primary
Clone 287219
Conjugate APC
Cross Reactivity Human
Antigen ILT4/CD85d
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human ILT4. Gly24-His458
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Monoclonal or Polyclonal Monoclonal