Please login to your online account to display your discounted pricing

anti-ILT4/CD85d, Alexa Fluor 700, Clone: 287219, R&D Systems


Manufacturer: r & d systems  RDSFAB2078N100

 View more versions of this product

Catalog No. FAB2078N100

  • / Each
 

Description & Specifications

Specifications

Clone 287219
Regulatory Status RUO
Cross Reactivity Human
Purification Method Protein A or G purified from hybridoma culture supernatant
Immunogen Mouse myeloma cell line NS0-derived recombinant human ILT4/CD85d. Gly24-His458
Host Species Mouse
Format Purified
Conjugate Alexa Fluor 700
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Antigen ILT4/CD85d
Isotype IgG2a
Quantity 100 Tests