Please login to your online account to display your discounted pricing

anti-IL1RL1, Polyclonal, Novus Biologicals™

$139.00 - $307.00

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A synthetic peptide corresponding to amino acids 207-238 of human ST2V was used as immunogen
Antigen IL1RL1
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224706SS

 
novus biologicals canada
NBP224706SS
Unlabeled 0.025mg Each for $139.00

NBP224706

 
novus biologicals canada
NBP224706
Unlabeled 0.1mg Each for $307.00
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A synthetic peptide corresponding to amino acids 207-238 of human ST2V was used as immunogen
Antigen IL1RL1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform1 and isoform 2), monkey and rat, and 91% homologous in chicken.
Gene Accession No. Q01638
Gene ID (Entrez) 9173
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL