Please login to your online account to display your discounted pricing

anti-IL-7 R alpha/CD127, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP229618

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP229618

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 3575
Formulation PBS
Gene Accession No. P16871
Concentration 0.5mg/mL
Antigen IL-7 R alpha/CD127
Applications Western Blotting
Primary or Secondary Primary
Quantity 0.1mg
Immunogen Partial recombinant protein made to an internal portion of human CRLF2 protein (between residues 100-300) [NCBI AF338733_1]
Regulatory Status RUO
Format Purified
Cross Reactivity Human
Conjugate Unlabeled
Host Species Rabbit