Please login to your online account to display your discounted pricing

anti-IL-7 R alpha/CD127, Polyclonal, Novus Biologicals™

$109.00 - $219.00

Specifications

Host Species Rabbit
Immunogen Partial recombinant protein made to an internal portion of human CRLF2 protein (between residues 100-300) [NCBI AF338733_1]
Antigen IL-7 R alpha/CD127
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP229618

 
novus biologicals canada
NBP229618
Unlabeled 0.1mg Each for $219.00

NBP229618SS

 
novus biologicals canada
NBP229618SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Partial recombinant protein made to an internal portion of human CRLF2 protein (between residues 100-300) [NCBI AF338733_1]
Antigen IL-7 R alpha/CD127
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Gene Accession No. P16871
Gene ID (Entrez) 3575
Formulation PBS
Concentration 0.5mg/mL