Please login to your online account to display your discounted pricing

anti-IL-7 R alpha/CD127, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP229618SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP229618SS

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Regulatory Status RUO
Concentration 0.5mg/mL
Antigen IL-7 R alpha/CD127
Formulation PBS
Gene ID (Entrez) 3575
Gene Accession No. P16871
Conjugate Unlabeled
Format Purified
Cross Reactivity Human
Immunogen Partial recombinant protein made to an internal portion of human CRLF2 protein (between residues 100-300) [NCBI AF338733_1]
Host Species Rabbit
Applications Western Blotting
Monoclonal or Polyclonal Polyclonal
Quantity 0.025mg