Please login to your online account to display your discounted pricing
anti-IL-4 Clone: BVD4-1D11, Thermo Scientific™ Novex™ DFS Item
IL-4 Monoclonal antibody AMC0042 from Thermo Scientific™ Novex™ specifically detects IL-4 in Mouse samples, and it is validated for FACS, ELISA and Neu
$443.00 - $450.00
Products
Catalog Number Mfr. No. Price Quantity    
AMC0044
DFS Item
life technologies
LSAMC0044
Each for $443.00
AMC0042
DFS Item
life technologies
LSAMC0042
Each for $450.00
Description & Specifications
The protein encoded by this gene is a pleiotropic cytokine produced by activated T cells. This cytokine is a ligand for interleukin 4 receptor. The interleukin 4 receptor also binds to IL13, which may contribute to many overlapping functions of this cytokine and IL13. STAT6, a signal transducer and activator of transcription, has been shown to play a central role in mediating the immune regulatory signal of this cytokine. This gene, IL3, IL5, IL13, and CSF2 form a cytokine gene cluster on chromosome 5q, with this gene particularly close to IL13. This gene, IL13 and IL5 are found to be regulated coordinately by several long-range regulatory elements in an over 120 kilobase range on the chromosome. Two alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported.
AMC0042 detects IL-4 in Mouse samples. AMC0042 has been successfully used in FACS, ELISA and Neu procedures. The immunogen for AMC0042 is recombinant mouse IL-4.
Applications:
ELISA, FACS
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.