Please login to your online account to display your discounted pricing

anti-IL-32, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224728

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224728

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 9235
Formulation 100μg in 200μL PBS containing 0.05% BSA
Applications Western Blotting
Gene Accession No. P24001
Concentration 0.5mg/mL
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_b) and chimpanzee, 93% homologous in horse and 87% homologous in cow, mouse and rat.
Cross Reactivity Human
Antigen IL-32
Immunogen A portion of amino acid 200-234 of human IL-32 was used as the immunogen.
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Isotype IgG
Regulatory Status RUO
Host Species Rabbit
Conjugate Unlabeled
Format Purified
Quantity 0.1mg