missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-IL-32, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Isotype IgG
Immunogen The amino acid sequence PELTPLLEKERDGLRC was used as the immunogen
Antigen IL-32
Monoclonal or Polyclonal Polyclonal
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224774SS Novus Biologicals Canada
NBP224774SS
0.025mg Each for $109.00
NBP224774 Novus Biologicals Canada
NBP224774
0.1mg Each for $255.80
Specifications Description & Specifications

Specifications

Rabbit
The amino acid sequence PELTPLLEKERDGLRC was used as the immunogen
Polyclonal
Primary
ELISA
P24001
100μg in 200μL PBS containing 0.2% gelatin
IgG
IL-32
Purified
RUO
The amino acid sequence used is 83% homologous in mouse and rat.
9235
0.5mg/mL