Please login to your online account to display your discounted pricing

anti-IL-32, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acid 200-234 of human IL-32 was used as the immunogen.
Antigen IL-32
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224728SS

 
novus biologicals canada
NBP224728SS
Unlabeled 0.025mg Each for $109.00

NBP224728

 
novus biologicals canada
NBP224728
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acid 200-234 of human IL-32 was used as the immunogen.
Antigen IL-32
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_b) and chimpanzee, 93% homologous in horse and 87% homologous in cow, mouse and rat.
Gene Accession No. P24001
Gene ID (Entrez) 9235
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL