Please login to your online account to display your discounted pricing

anti-IL-32, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224774SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224774SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 9235
Concentration 0.5mg/mL
Gene Accession No. P24001
Primary or Secondary Primary
Quantity 0.025mg
Conjugate Unlabeled
Formulation 100μg in 200μL PBS containing 0.2% gelatin
Test Specificity The amino acid sequence used is 83% homologous in mouse and rat.
Isotype IgG
Regulatory Status RUO
Format Purified
Immunogen The amino acid sequence PELTPLLEKERDGLRC was used as the immunogen
Applications ELISA
Cross Reactivity Human
Antigen IL-32
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit