Please login to your online account to display your discounted pricing

anti-IL-32, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224774

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224774

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg in 200μL PBS containing 0.2% gelatin
Gene Accession No. P24001
Gene ID (Entrez) 9235
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Primary or Secondary Primary
Test Specificity The amino acid sequence used is 83% homologous in mouse and rat.
Concentration 0.5mg/mL
Immunogen The amino acid sequence PELTPLLEKERDGLRC was used as the immunogen
Antigen IL-32
Format Purified
Regulatory Status RUO
Applications ELISA
Host Species Rabbit
Isotype IgG
Quantity 0.1mg
Cross Reactivity Human