Please login to your online account to display your discounted pricing

IL-3 Polyclonal Antibody, Invitrogen™


IL-3 Antibody, PA1-75208, from Invitrogen™. Species Reactivity: Human; Applications: ELISA.

Manufacturer: pierce biotechnology inc  PA175208

Catalog No. PIPA175208

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Formulation 500μg purified antibody in PBS and 0.09% sodium azide
Gene ID (Entrez) 3562
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Gene Accession No. P08700
Gene Alias IL-3, MCGF, MGC79398, MGC79399, MULTI-CSF
Species Reactivity Human
Isotype IgG
Storage Requirements Avoid Freeze/Thaw Cycles
Format Purified
Quantity 500μg
Immunogen Recombinant human IL-3
Antigen IL-3
Regulatory Status RUO
Cross Reactivity Human
Host Species Sheep
Classification Polyclonal
Gene Symbols IL3
Conjugate Unlabeled
Type Primary

IL-3 Antibody, PA1-75208, from Invitrogen. Species Reactivity: Human; Applications: ELISA.

The protein encoded by this gene is a potent growth promoting cytokine. This cytokine is capable of supporting the proliferation of a broad range of hematopoietic cell types. It is involved in a variety of cell activities such as cell growth, differentiation and apoptosis. This cytokine has been shown to also possess neurotrophic activity, and it may be associated with neurologic disorders.
Host Species: Sheep
Cross Reactivity: Human
Isotype: IgG
Immunogen: Recombinant human IL-3
Formulation: 500μg purified antibody in PBS and 0.09% sodium azide
Synonyms: IL-3, MCGF, MGC79398, MGC79399, MULTI-CSF
Uniprot Accession: P08700
Entrez GeneID: 3562
Gene Symbol: IL3
PA1-75208 specifically detects IL-3. PA1-75208 has been successfully used in ELISA applications. The PA1-75208 immunogen is Recombinant IL-3.