Please login to your online account to display your discounted pricing

anti-IL-3 Polyclonal, Thermo Scientific™

IL-3 Polyclonal antibody PA1-75208 from Thermo Scientific™ specifically detects IL-3 in Human samples, and it is validated for ELISA

$2,030.00 - $2,030.00

Specifications

Antigen IL-3
Applications Enzyme Linked Immunosorbent Assay
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

PIPA175208

 
pierce biotechnology inc
PA175208
Each for $2,030.00
Description & Specifications

Specifications

Antigen IL-3
Applications Enzyme Linked Immunosorbent Assay
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Host Species Sheep
Quantity 500μg
Regulatory Status RUO
Type Primary
Formulation 500μg purified antibody in PBS and 0.09% sodium azide
Gene Symbols IL3
Immunogen Recombinant human IL-3
Storage Requirements Avoid Freeze/Thaw Cycles

IL-3 Polyclonal antibody PA1-75208 from Thermo Scientific™ specifically detects IL-3 in Human samples, and it is validated for ELISA

The protein encoded by this gene is a potent growth promoting cytokine. This cytokine is capable of supporting the proliferation of a broad range of hematopoietic cell types. It is involved in a variety of cell activities such as cell growth, differentiation and apoptosis. This cytokine has been shown to also possess neurotrophic activity, and it may be associated with neurologic disorders.
Host Species: Sheep
Cross Reactivity: Human
Isotype: IgG
Immunogen: Recombinant human IL-3
Formulation: 500μg purified Antibody in PBS, with 0.09% sodium azide;
Synonyms: IL-3, MCGF, MGC79398, MGC79399, MULTI-CSF
Uniprot Accession: P08700
Gene Symbol: IL3
Entrez GeneID: 3562
PA1-75208 detects IL-3 in Human samples. PA1-75208 has been successfully used in ELISA procedures. PA1-75208 The immunogen is Recombinant human IL-3.