Please login to your online account to display your discounted pricing

anti-IL-27, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 31-46 (RPPGRPQLSLQELRRE) of human IL-27 (p28 subunit) of Genbank accession number NP_663634 was used as immunogen.
Antigen IL-27
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056389

 
novus biologicals canada
NB10056389
Unlabeled 0.1mg Each for $255.80

NB10056389T

 
novus biologicals canada
NB10056389SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 31-46 (RPPGRPQLSLQELRRE) of human IL-27 (p28 subunit) of Genbank accession number NP_663634 was used as immunogen.
Antigen IL-27
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Gene Accession No. Q8NEV9
Gene ID (Entrez) 246778
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL