Please login to your online account to display your discounted pricing

anti-IL-27/IL-35 EBI3 Subunit, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP203935SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP203935SS

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Gene ID (Entrez) 10148
Conjugate Unlabeled
Test Specificity EBI3 is routinely seen in western blot at approximately 35 kD. The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a and CRA_b), Reacts with Chimpanzee and Human. Predicted to cross react with New World Monkey.
Cross Reactivity Human
Cross Reactivity Primate
Gene Accession No. Q14213
Formulation 100μg in 200μL PBS containing 0.05% BSA
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal
Immunogen A portion of amino acids 120-170 of human EBI3 was used as the immunogen
Antigen IL-27/IL-35 EBI3 Subunit
Format Purified
Host Species Rabbit
Applications Western Blotting
Quantity 0.025mg