Please login to your online account to display your discounted pricing

anti-IL-27/IL-35 EBI3 Subunit, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP203935

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP203935

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 10148
Gene Accession No. Q14213
Conjugate Unlabeled
Quantity 0.1mg
Formulation 100μg in 200μL PBS containing 0.05% BSA
Test Specificity EBI3 is routinely seen in western blot at approximately 35 kD. The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a and CRA_b), Reacts with Chimpanzee and Human. Predicted to cross react with New World Monkey.
Cross Reactivity Human
Cross Reactivity Primate
Immunogen A portion of amino acids 120-170 of human EBI3 was used as the immunogen
Antigen IL-27/IL-35 EBI3 Subunit
Format Purified
Regulatory Status RUO
Primary or Secondary Primary
Host Species Rabbit
Applications Western Blotting