Please login to your online account to display your discounted pricing

anti-IL-27/IL-35 EBI3 Subunit, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227359SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227359SS

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Host Species Rabbit
Gene ID (Entrez) 10148
Conjugate Unlabeled
Concentration 0.5mg/mL
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in mouse and rat and 85% homologous in human.
Primary or Secondary Primary
Format Purified
Immunogen Amino acids 51-77 of mouse EBI3 were used as the immunogen for this antibody.
Quantity 0.025mg
Cross Reactivity Mouse
Applications Western Blotting
Regulatory Status RUO
Antigen IL-27/IL-35 EBI3 Subunit