Please login to your online account to display your discounted pricing

anti-IL-27/IL-35 EBI3 Subunit, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227359

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227359

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Concentration 0.5mg/mL
Applications Western Blotting
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Gene ID (Entrez) 10148
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in mouse and rat and 85% homologous in human.
Host Species Rabbit
Antigen IL-27/IL-35 EBI3 Subunit
Regulatory Status RUO
Immunogen Amino acids 51-77 of mouse EBI3 were used as the immunogen for this antibody.
Quantity 0.1mg
Cross Reactivity Mouse
Primary or Secondary Primary
Format Purified