Please login to your online account to display your discounted pricing

anti-IL-27/IL-35 EBI3 Subunit, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP203937SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP203937SS

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg in 200μL PBS containing 0.05% BSA
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Gene Accession No. Q14213
Gene ID (Entrez) 10148
Primary or Secondary Primary
Regulatory Status RUO
Cross Reactivity Human
Cross Reactivity Mouse
Host Species Rabbit
Format Purified
Applications Western Blotting
Quantity 0.025mg
Antigen IL-27/IL-35 EBI3 Subunit
Immunogen A portion of amino acids 50-100 of human EBI3 was used as the immunogen.