Please login to your online account to display your discounted pricing

anti-IL-27/IL-35 EBI3 Subunit, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP203937

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP203937

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. Q14213
Host Species Rabbit
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Formulation 100μg in 200μL PBS containing 0.05% BSA
Gene ID (Entrez) 10148
Quantity 0.1mg
Immunogen A portion of amino acids 50-100 of human EBI3 was used as the immunogen.
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Antigen IL-27/IL-35 EBI3 Subunit
Primary or Secondary Primary
Applications Western Blotting
Regulatory Status RUO