missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-IL-27/IL-35 EBI3 Subunit, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen Amino acids 51-77 of mouse EBI3 were used as the immunogen for this antibody.
Antigen IL-27/IL-35 EBI3 Subunit
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP227359 Novus Biologicals Canada
NBP227359
0.1mg Each for $255.80
NBP227359SS Novus Biologicals Canada
NBP227359SS
0.025mg Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
IL-27/IL-35 EBI3 Subunit
Purified
RUO
The amino acid sequence used as immunogen for this antibody is 100% homologous in mouse and rat and 85% homologous in human.
100μg antibody in 200μL PBS containing 0.05% BSA.
Amino acids 51-77 of mouse EBI3 were used as the immunogen for this antibody.
Polyclonal
Primary
Western Blot
10148
0.5mg/mL