missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-IL-27/IL-35 EBI3 Subunit, Polyclonal, Novus Biologicals™

$139.00 - $307.00

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 50-100 of human EBI3 was used as the immunogen.
Antigen IL-27/IL-35 EBI3 Subunit
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP203937SS Novus Biologicals Canada
NBP203937SS
0.025mg Each for $139.00
NBP203937 Novus Biologicals Canada
NBP203937
0.1mg Each for $307.00
Specifications Description & Specifications

Specifications

Rabbit
IL-27/IL-35 EBI3 Subunit
Purified
RUO
Q14213
100μg in 200μL PBS containing 0.05% BSA
A portion of amino acids 50-100 of human EBI3 was used as the immunogen.
Polyclonal
Primary
Western Blot
10148