missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-IL-27/IL-35 EBI3 Subunit, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 120-170 of human EBI3 was used as the immunogen
Antigen IL-27/IL-35 EBI3 Subunit
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP203935 Novus Biologicals Canada
NBP203935
0.1mg Each for $255.80
NBP203935SS Novus Biologicals Canada
NBP203935SS
0.025mg Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
IL-27/IL-35 EBI3 Subunit
Purified
RUO
EBI3 is routinely seen in western blot at approximately 35 kD. The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a and CRA_b), Reacts with Chimpanzee and Human. Predicted to cross react with New World Monkey.
10148
A portion of amino acids 120-170 of human EBI3 was used as the immunogen
Polyclonal
Primary
Western Blot
Q14213
100μg in 200μL PBS containing 0.05% BSA