Please login to your online account to display your discounted pricing

anti-IL-24, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP223906SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP223906SS

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Formulation 100μL of crude rabbit sera with 50% glycerol
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 11009
Cross Reactivity Human
Immunogen Within the range of amino acids 159-194 of human IL-24 protein
Format Antiserum
Regulatory Status RUO
Applications Western Blotting
Host Species Rabbit
Conjugate Unlabeled
Quantity 0.025mL
Antigen IL-24