Please login to your online account to display your discounted pricing

anti-IL-24, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP223906

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP223906

  • / Each
 

Description & Specifications

Specifications

Antigen IL-24
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Human
Format Antisera
Formulation 100μL of crude rabbit sera with 50% glycerol
Host Species Rabbit
Immunogen Within the range of amino acids 159-194 of human IL-24 protein
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 11009
Target Species Human