Please login to your online account to display your discounted pricing

anti-IL-24, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 159-194 of human IL-24 protein
Antigen IL-24
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP223906

 
novus biologicals canada
NBP223906
Unlabeled 0.1mL Each for $255.80

NBP223906SS

 
novus biologicals canada
NBP223906SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 159-194 of human IL-24 protein
Antigen IL-24
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Gene ID (Entrez) 11009
Formulation 100μL of crude rabbit sera with 50% glycerol