Please login to your online account to display your discounted pricing

anti-IL-18 R beta/IL-1 R7, PE, Clone: 132029, R&D Systems


Manufacturer: r & d systems  FAB118P025

 View more versions of this product

Catalog No. FAB118P025

  • / Each
 

Description & Specifications

Specifications

Immunogen Met1-Arg356
Clone 132029
Format Purified
Quantity 25 Tests
Antigen IL-18 R beta/IL-1 R7
Regulatory Status RUO
Host Species Mouse
Applications Flow Cytometry
Isotype IgG2b
Cross Reactivity Human
Purification Method Protein A or G purified from hybridoma culture supernatant
Formulation Supplied in a saline solution containing BSA and Sodium Azide.
Test Specificity Detects human IL‑18 Rβ/IL‑1 R7 in direct ELISAs and Western blots. In direct ELISAs, 15% cross-reactivity with recombinant human (rh) IL‑1 RII is observed and 2% cross-reactivity with rhIL-1 R9 is observed.
Conjugate PE
Primary or Secondary Primary
Gene ID (Entrez) 8807
Gene Accession No. O95256
Monoclonal or Polyclonal Monoclonal