Please login to your online account to display your discounted pricing

anti-IL-17B, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056892

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056892

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Test Specificity In HeLa, a 45 kDa band is observed.
Concentration 0.5mg/mL
Formulation PBS containing 0.1mg/mL BSA
Gene Accession No. Q9UHF5
Conjugate Unlabeled
Antigen IL-17B
Format Purified
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 27190
Regulatory Status RUO
Applications Western Blotting
Immunogen Amino acids 102-118 (CNYTEAFQTQTRPSGGK) of human IL-17BR protein were used as the immunogen.
Host Species Rabbit
Cross Reactivity Human
Quantity 0.2mL