Please login to your online account to display your discounted pricing

anti-IL-17B, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen Amino acids 102-118 (CNYTEAFQTQTRPSGGK) of human IL-17BR protein were used as the immunogen.
Antigen IL-17B
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056892T

 
novus biologicals canada
NB10056892SS
Unlabeled 0.05mL Each for $139.00

NB10056892

 
novus biologicals canada
NB10056892
Unlabeled 0.2mL Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Amino acids 102-118 (CNYTEAFQTQTRPSGGK) of human IL-17BR protein were used as the immunogen.
Antigen IL-17B
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity In HeLa, a 45 kDa band is observed.
Gene Accession No. Q9UHF5
Gene ID (Entrez) 27190
Formulation PBS containing 0.1mg/mL BSA
Concentration 0.5mg/mL