Please login to your online account to display your discounted pricing

anti-IL-1 beta/IL-1F2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP119775

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP119775

  • / Each
 

Description & Specifications

Specifications

Formulation PBS
Gene Accession No. P01584
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Antigen IL-1 beta/IL-1F2
Format Affinity Purified
Gene ID (Entrez) 3553
Concentration 1.0mg/mL
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Quantity 0.1mg
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Immunogen Synthetic peptide made to a C-terminal portion of the human IL 1 beta protein (between amino acids 100-200) [UniProt P01584]
Host Species Rabbit