Please login to your online account to display your discounted pricing

anti-IL-1 beta/IL-1F2, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to a C-terminal portion of the human IL 1 beta protein (between amino acids 100-200) [UniProt P01584]
Antigen IL-1 beta/IL-1F2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP119775

 
novus biologicals canada
NBP119775
Unlabeled 0.1mg Each for $309.05

NBP119775SS

 
novus biologicals canada
NBP119775SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to a C-terminal portion of the human IL 1 beta protein (between amino acids 100-200) [UniProt P01584]
Antigen IL-1 beta/IL-1F2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Gene Accession No. P01584
Gene ID (Entrez) 3553
Formulation PBS
Concentration 1.0mg/mL