Please login to your online account to display your discounted pricing

anti-IKZF3, Polyclonal, Novus Biologicals™

$109.00 - $362.00

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 350-400 of human IKZF3 was used as the immunogen.
Antigen IKZF3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224495

 
novus biologicals canada
NBP224495
Unlabeled 0.1mg Each for $362.00

NBP224495SS

 
novus biologicals canada
NBP224495SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 350-400 of human IKZF3 was used as the immunogen.
Antigen IKZF3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee and rhesus monkey, 88% in dog, mouse and rat and 82% homologous in horse.
Gene Accession No. Q9UKT9
Gene ID (Entrez) 22806
Formulation PBS
Concentration 0.5mg/mL