missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-IKB zeta, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Immunogen Synthetic peptides corresponding to an amino acid range between 270-320 and 670-720 of mouse IkB zeta protein were used as the immunogen for this antibody.
Antigen IKB zeta
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB10056390 Novus Biologicals Canada
NB10056390
0.1mg Each for $259.02
NB10056390T Novus Biologicals Canada
NB10056390SS
0.025mg Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
IKB zeta
Purified
RUO
This antibody is reactive against Mouse. Predicted cross-reactivity based on sequence identity: Mouse (100%), Human, (100%), Rat (100%), Chimp (100%), Bovine (100%), Chicken (93%). Not yet tested in other species.
64332
0.5mg/mL
Synthetic peptides corresponding to an amino acid range between 270-320 and 670-720 of mouse IkB zeta protein were used as the immunogen for this antibody.
Polyclonal
Primary
Western Blot
Q9BYH8
PBS containing 0.05% BSA