Please login to your online account to display your discounted pricing

anti-IkB-alpha, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP223901SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP223901SS

  • / Each
 

Description & Specifications

Specifications

Antigen IkB-alpha
Applications Peptide ELISA
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Human
Format Antisera
Formulation 100μL; IkB alpha anti-sera with 50% glycerol
Gene Accession No. P25963
Host Species Rabbit
Immunogen Within the range of amino acids 130-164 of human IkB alpha protein were used as the immunogen.
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 4792
Target Species Human