Please login to your online account to display your discounted pricing

anti-IkB-alpha, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP223901

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP223901

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 4792
Formulation 100μL; IkB alpha anti-sera with 50% glycerol
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Antigen IkB-alpha
Gene Accession No. P25963
Applications Peptide ELISA
Applications Western Blotting
Quantity 0.1mL
Cross Reactivity Human
Format Antiserum
Host Species Rabbit
Regulatory Status RUO
Immunogen Within the range of amino acids 130-164 of human IkB alpha protein were used as the immunogen.