Please login to your online account to display your discounted pricing

anti-IkB-alpha, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056416SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056416T

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 4792
Gene Accession No. P25963
Conjugate Unlabeled
Primary or Secondary Primary
Formulation PBS containing 0.2% gelatin
Test Specificity This specific epitope of this has not been mapped, but it does not appear to detect mouse IkBa.
Antigen IkB-alpha
Concentration 0.5mg/mL
Regulatory Status RUO
Applications Western Blotting
Quantity 0.025mg
Immunogen A full-length recombinant protein of human IkBa was used as immunogen.
Host Species Rabbit
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Human
Format Purified