Please login to your online account to display your discounted pricing

anti-IkB-alpha, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056416

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056416

  • / Each
 

Description & Specifications

Specifications

Antigen IkB-alpha
Conjugate Unlabeled
Gene ID (Entrez) 4792
Test Specificity This specific epitope of this has not been mapped, but it does not appear to detect mouse IkBa.
Formulation PBS containing 0.2% gelatin
Gene Accession No. P25963
Concentration 0.5mg/mL
Regulatory Status RUO
Host Species Rabbit
Cross Reactivity Human
Format Purified
Applications Western Blotting
Primary or Secondary Primary
Quantity 0.1mg
Monoclonal or Polyclonal Polyclonal
Immunogen A full-length recombinant protein of human IkBa was used as immunogen.