Please login to your online account to display your discounted pricing

anti-IkB-alpha, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A full-length recombinant protein of human IkBa was used as immunogen.
Antigen IkB-alpha
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056416T

 
novus biologicals canada
NB10056416SS
Unlabeled 0.025mg Each for $139.00

NB10056416

 
novus biologicals canada
NB10056416
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A full-length recombinant protein of human IkBa was used as immunogen.
Antigen IkB-alpha
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity This specific epitope of this has not been mapped, but it does not appear to detect mouse IkBa.
Gene Accession No. P25963
Gene ID (Entrez) 4792
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL