Please login to your online account to display your discounted pricing

anti-IGF-I R, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP177679SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP177679SS

  • / Each
 

Description & Specifications

Specifications

Antigen IGF-I R
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blot
Concentration 1.05mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Affinity Purified
Formulation PBS, 30% glycerol
Gene Accession No. P08069
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human IGF1 Receptor protein (between residues 1300-1367) [UniProt P08069].
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 3480
Target Species Human
Target Species Murine
Target Species Primate
Target Species Rat