missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-IGF-I R, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human IGF1 Receptor protein (between residues 700-800) [UniProt P08069]
Antigen IGF-I R
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP177680SS Novus Biologicals Canada
NBP177680SS
0.025mL Each for $139.00
NBP177680 Novus Biologicals Canada
NBP177680
0.1mL Each for $309.05
Specifications Description & Specifications

Specifications

Rabbit
IGF-I R
Affinity Purified
RUO
Human, rat, mouse and primate. Reactivity with Equine reported via customer feedback. Predicted to react with Bovine (100%).
3480
1.09mg/mL
A synthetic peptide made to an internal portion of the human IGF1 Receptor protein (between residues 700-800) [UniProt P08069]
Polyclonal
Primary
Immunocytochemistry,Immunofluorescence,Western Blot
P08069
PBS, 30% glycerol